Adipic acid ≥99%

Warning

Synonyms: Hexanedioc acid, 1,6-Hexanedioic acid

A26357-3KG A26357-1KG A26357-25G A26357-100G A26357-500G
220101-603EA 88.62 USD
220101-603 220101-601 220101-602 220102-398 220101-827
Adipic acid ≥99%
Adipic acid
Formula: HOOC(CH₂)₄COOH
MW: 146.14 g/mol
Boiling Pt: 337 °C (1013 hPa)
Melting Pt: 151 °C
Density: 1.36 g/cm³ (25 °C)
Flash Pt: 196 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004420
CAS Number: 124-04-9
EINECS: 204-673-3
Merck Index: 13,00163
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58ac39e8ebd69527d91cb2d15a469a2a69506785&vid=ef907d9f9fd34903274c42b99d1d1ac1a8db6dce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR