Acrolein diethyl acetal ≥96%

Synonyms: Acrolein Acetal, 1,1-Diethoxy-2-propene, 3,3-Diethoxy-1-propene

A24001-100G A24001-25G
220102-300EA 232.62 USD
220102-300 220102-301
Acrolein diethyl acetal ≥96%
Acrolein diethyl acetal
Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 125 °C (1013 hPa)
Melting Pt: –77 °C
Density: 0.8543 g/cm³ (15 °C)
MDL Number: MFCD00009239
CAS Number: 3054-95-3
EINECS: 221-276-0
UN: 2374
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR