2-Acetylthiophene

Danger

Synonyms: 1-Thiophen-2-yl-ethanone, 1-(2-Thienyl)-1-ethanone, Methyl 2-thienyl ketone

A22602-25G A22602-100G
220102-195EA 40.92 USD
220102-195 220102-194
2-Acetylthiophene
2-Acetylthiophene
Formula: C₆H₆OS
MW: 126.18 g/mol
Boiling Pt: 214 °C (1013 hPa)
Melting Pt: 9 °C
Density: 1.1704 g/cm³ (20 °C)
MDL Number: MFCD00005442
CAS Number: 88-15-3
EINECS: 201-804-6
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR