4-Acetylpyridine

Synonyms: Methyl 4-pyridyl ketone

A21401-100G A21401-10G
220102-135EA 142.15 USD
220102-135 220102-136
4-Acetylpyridine
4-Acetylpyridine
Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: 14 - 16 °C
Density: 1.1009 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006433
CAS Number: 1122-54-9
EINECS: 214-350-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR