Aluminium ammonium sulfate dodecahydrate, ReagentPlus®

Warning

A2140-500G A2140-1KG
220102-134EA 113.85 USD
220102-134 220102-133
Aluminium ammonium sulfate dodecahydrate, ReagentPlus®
Aluminium ammonium sulphate dodecahydrate
Formula: AlNH₄(SO₄)₂·12H₂O
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 93 °C
Density: 1.64 g/cm³ (20 °C)
MDL Number: MFCD00051042
CAS Number: 7784-26-1
EINECS: 232-055-3
Merck Index: 13,00325

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR