(S)-(+)-2-Aminobutyric acid ≥99%

A1879-1G A1879-5G
220101-587EA 141.54 USD
220101-587 220101-588
(S)-(+)-2-Aminobutyric acid ≥99%
(S)-(+)-2-Aminobutyric acid
Formula: C₄H₉NO₂
MDL Number: MFCD00064415
CAS Number: 1492-24-6
EINECS: 216-083-3

Order Now

Specification Test Results

Assay 99% (titration)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR