4-Aminohippuric acid ≥99%

Synonyms: p-Aminohippuric acid, 4-Aminohippuric Acid, 2-(4-Aminobenzamido)acetic acid, N-(4-Aminobenzoyl)glycine

A1422-25G A1422-10G A1422-250G A1422-100G
220101-815EA 256 USD
220101-815 220101-813 220101-814 220101-812
4-Aminohippuric acid ≥99%
4-Aminohippuric acid
Formula: H₂NC₆H₄C(O)NHCH₂COOH
MW: 194.19 g/mol
Boiling Pt: 441 °C (1013 hPa)
Melting Pt: 199 - 202 °C
Density: 1.402 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007890
CAS Number: 61-78-9
EINECS: 200-518-9
Merck Index: 13,00441

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR