Triammonium citrate ≥97%

Warning

Synonyms: 2-Hydroxy-1,2,3-propanetricarboxylic acid triammonium salt, Ammonium citrate tribasic, Citric acid triammonium salt, Triammonium 2-hydroxypropane-1,2,3-tricarboxylate

A1332-100G A1332-500G
220100-306EA 71.72 USD
220100-306 220100-307
Triammonium citrate ≥97%
Triammonium citrate
Formula: C₆H₁₇N₃O₇
MW: 243.22 g/mol
Melting Pt: 185 °C
Density: 1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036406
CAS Number: 3458-72-8
EINECS: 222-394-5

Order Now

Specification Test Results

Assay 97% (titration)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR