4'-Hydroxy-3'-methoxyacetophenone ≥98%

Synonyms: 4-Hydroxy-3-methoxy-acetophenone, 1-(4-Hydroxy-3-methoxyphenyl)ethanone, Acetovanillone, Apocynin

A10809-100G A10809-25G
220101-698EA 126 USD
220101-698 220101-699
4'-Hydroxy-3'-methoxyacetophenone ≥98%
4'-Hydroxy-3'-methoxyacetophenone
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 115 °C
MDL Number: MFCD00008747
CAS Number: 498-02-2
EINECS: 207-854-5
Merck Index: 13,00746

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR