DL-Valine ≥99%

94640-25G 94640-500G 94640-100G
220100-610EA 50.08 USD
220100-610 220100-611 220100-609
DL-Valine ≥99%
DL-Valine
Formula: C₅H₁₁NO₂
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 86 °C
Density: 1.352 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004267
CAS Number: 516-06-3
EINECS: 208-220-0
Merck Index: 12,10046

Order Now

Specification Test Results

Assay 99% (NT)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR