1-Butyl-3-methylimidazolium chloride

Synonyms: 3-Butyl-1-methyl-1H-imidazol-3-ium chloride

94128-50G 94128-5G 94128-250G
220099-596EA 175 USD
220099-596 220099-597 220099-595
1-Butyl-3-methylimidazolium chloride
1-Butyl-3-methylimidazolium chloride
Formula: C₈H₁₅ClN₂
MW: 174.67 g/mol
Boiling Pt: ∼200 °C (1013 hPa)
Melting Pt: ∼60 °C
Density: ~ 1 g/cm³ (20 °C)
Flash Pt: >110 °C
MDL Number: MFCD03095425
CAS Number: 79917-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR