Triphenylphosphine

Warning

93092-1KG 93092-250G
220098-254EA 149.23 USD
220098-254 220098-255
Triphenylphosphine
Triphenylphosphine
Formula: C₁₈H₁₅P
MW: 262.29 g/mol
Boiling Pt: 377 °C (1013 hPa)
Melting Pt: 79 °C
Density: 1.194 g/cm³ (20 °C)
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00003043
CAS Number: 603-35-0
EINECS: 210-036-0
Merck Index: 12,09873

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR