2',4',6'-Trimethylacetophenone

Synonyms: 1-(2,4,6-Trimethylphenyl)ethan-1-one

92220-10ML 92220-50ML
220100-069EA 84 USD
220100-069 220100-070
2',4',6'-Trimethylacetophenone
2',4',6'-Trimethylacetophenone
Formula: C₁₁H₁₄O
MW: 162.23 g/mol
Boiling Pt: 240…248 °C (1013 hPa)
Density: 0.975 g/cm³ (20 °C)
MDL Number: MFCD00008735
CAS Number: 1667-01-2
EINECS: 216-783-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR