3,4,5-Trimethoxybenzyl chloride

Danger

Synonyms: 3,4,5-Trimethoxybenzylchloride, 5-(Chloromethyl)-1,2,3-trimethoxybenzene

92176-5G 92176-25G
220100-061EA 115.23 USD
220100-061 220098-101
3,4,5-Trimethoxybenzyl chloride
3,4,5-Trimethoxybenzyl chloride
Formula: C₁₀H₁₃ClO₃
MW: 216.66 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00040716
CAS Number: 3840-30-0
EINECS: 223-330-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR