(3,3,3-Trifluoropropyl)trimethoxysilane

Synonyms: Trimethoxy(3,3,3-trifluoropropyl)silane

91877-25ML 91877-5ML
220100-009EA 206.31 USD
220100-009 220100-010
(3,3,3-Trifluoropropyl)trimethoxysilane
(3,3,3-Trifluoropropyl)trimethoxysilane
Formula: F₃CCH₂CH₂Si(OCH₃)₃
MW: 218.25 g/mol
Boiling Pt: 144 °C (1013 hPa)
Density: 1.137 g/cm³ (20 °C)
MDL Number: MFCD00039266
CAS Number: 429-60-7
EINECS: 207-059-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR