1-Hexyl-3-methylimidazolium hexafluorophosphate ≥97.0% (by HPLC), Sigma-Aldrich®

Synonyms: 1-Hexyl-3-methyl-1H-imidazol-3-ium hexafluorophosphate(V)

89320-50G 89320-5G
220099-323EA 353 USD
220099-323 220099-324
1-Hexyl-3-methylimidazolium hexafluorophosphate ≥97.0% (by HPLC), Sigma-Aldrich®
1-Hexyl-3-methylimidazolium hexafluorophosphate
Formula: C₁₀H₁₉F₆N₂P
MW: 312.24 g/mol
Density: 1.28 g/cm³ (20 °C)
MDL Number: MFCD03093296
CAS Number: 304680-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR