Titanium(IV) isopropoxide

87560-500ML 87560-100ML
220098-956EA 87.23 USD
220098-956 220098-955
Titanium(IV) isopropoxide
Titanium(IV) isopropoxide
Formula: C₁₂H₂₈O₄Ti
Boiling Pt: 240 °C (1013 hPa)
Melting Pt: 18 - 20 °C
Density: 0.9711 g/cm³ (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00008871
CAS Number: 546-68-9
EINECS: 208-909-6
UN: 2413
ADR: 3,III
Merck Index: 12,09614

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR