Tetrabutylammonium fluoride trihydrate

86872-10G 86872-250G 86872-50G
220098-800EA 87.77 USD
220098-800 220098-801 220098-802
Tetrabutylammonium fluoride trihydrate
Tetrabutylammonium fluoride trihydrate
Formula: C₁₆H₃₆FN·3H₂O
MW: 315.51 g/mol
Melting Pt: 62…63 °C
MDL Number: MFCD00149981
CAS Number: 87749-50-6
EINECS: 207-057-2
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR