Tetrabutylammonium chloride

Warning

86870-500G 86870-25G 86870-100G
220098-799EA 650.77 USD
220098-799 220098-798 220098-797
Tetrabutylammonium chloride
Tetrabutylammonium chloride
Formula: C₁₆H₃₆ClN
MW: 277.92 g/mol
Melting Pt: 83…86 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00011635
CAS Number: 1112-67-0
EINECS: 214-195-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR