2-Methyl-2-butene

Danger

Synonyms: Trimethylethylene, Isoamylene, Isopentene

86262-1L-F 86262-250ML-F 86262-50ML-F
220098-506EA 448 USD
220098-506 220098-507 220098-508
2-Methyl-2-butene
2-Methyl-2-butene
Formula: C₅H₁₀
MW: 70.13 g/mol
Boiling Pt: 39 °C (1013 hPa)
Melting Pt: –124 °C
Density: 0.6623 g/cm³ (20 °C)
Flash Pt: –45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009276
CAS Number: 513-35-9
EINECS: 208-156-3
UN: 2460
ADR: 3,II
Merck Index: 12,00649

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR