Thymol blue sodium salt ACS

Synonyms: Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bis[5-methyl-2-(1-methylethyl)phenolate] S,S-dioxide, Sodium 4-(3-(4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-2-isopropyl-5-methylphenolate, Thymol blue water soluble, Thymol Blue sodium

861367-25G 861367-5G
220098-443EA 173.6 USD
220098-443 220098-444
Thymol blue sodium salt ACS
Thymol blue sodium salt
Formula: C₂₇H₂₉NaO₅S
MW: 488.58 g/mol
Melting Pt: 283 °C
Density: 1.19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151093
CAS Number: 62625-21-2
EINECS: 263-650-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ebfc1bdaa6d2db4172c2e91dedebefd5e90e822&vid=55d8e4e97a25fd218390c34f3bc7e47526218e8f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR