Orange G, certified by the Biological Stain Commission

Synonyms: CI 16230

861286-100G 861286-25G
220098-437EA 382.2 USD
220098-437 220098-438
Orange G, certified by the Biological Stain Commission
Orange G
Formula: C₁₆H₁₀N₂Na₂O₇S₂
MW: 452.38 g/mol
Melting Pt: 141 °C
Density: 0.45 to 0.47 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012457
CAS Number: 1936-15-8
EINECS: 217-705-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR