Bismarck brown Y, certified by the Biological Stain Commission

Synonyms: C.I. 21000, 1,3-Bis(2,3-diaminophenylazo)benzene dihydrochloride, Bismarck Brown Y (G), Basic Brown 1, Vesuvine, Phenylene Brown, Leather Brown, Basic Brown G, GX, GXP, Manchester Brown, 4,4'-[1,3-Phenylenebis(azo)]bisbenzene-1,3-diamine dihydrochloride

861111-25G 861111-100G
220098-432EA 91.85 USD
220098-432 220098-431
Bismarck brown Y, certified by the Biological Stain Commission
Bismarck brown Y
Formula: C₁₈H₁₈N8·2HCl
MW: 419.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012977
CAS Number: 10114-58-6
EINECS: 233-314-3
Merck Index: 12,01294
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=277f38540157ea9c963d70dc2ceba7f9b8c2d493&vid=2415bd239294426b569d2b27ce4c8e9c3bdb4afc&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR