Eosin B, certified by the Biological Stain Commission

Synonyms: Eosin bluish, 2,7-Dinitro-4,5-dibromofluorescein, Eosin B disodium salt, Sodium 2-(4,5-dibromo-2,7-dinitro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate, 2-(4,5-dibromo-3,6-dihydroxy-2,7-dinitroxanthen-9-yl)-benzoic acid disodium salt, Acid Red 91

861006-25G 861006-100G 861006-10G
220098-422EA 214.2 USD
220098-422 220098-420 220098-421
Eosin B, certified by the Biological Stain Commission
Eosin B
Formula: C₂₀H₆Br₂N₂Na₂O₉
MW: 624.06 g/mol
Melting Pt: > 310 °C
Storage Temperature: Ambient
MDL Number: MFCD00005041
CAS Number: 548-24-3
EINECS: 208-943-1
Merck Index: 12,03638
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b44ed31611ccd8273c2aa49e942783c769935dd&vid=870721605fb3971a6b7131ac42941e4b53d2f762&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR