D-(+)-Proline

Synonyms: H-D-Pro-OH, (R)-(+)-Proline, D-Proline

858919-500MG 858919-5G
220098-369EA 48.35 USD
220098-369 220098-370
D-(+)-Proline
D-(+)-Proline
Formula: C₅H₉NO₂
MW: 115.13 g/mol
Melting Pt: 223 °C
MDL Number: MFCD00064317
CAS Number: 344-25-2
EINECS: 206-452-7

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR