Eriochrome Black T ACS indicator

Warning

Synonyms: 1-Naphthalenesulphonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1), Eriochrome black T, Indicator, EBT, Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulphonate, Solochrome Black T

858390-100G 858390-1KG 858390-500G
220097-372EA 44.52 USD
220097-372 220097-373 220097-374
Eriochrome Black T ACS indicator
Eriochrome Black T
Formula: C₂₀H₁₂N₃NaO₇S
MW: 461.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003935
CAS Number: 1787-61-7
EINECS: 217-250-3
UN: 3077
ADR: 9,III
Merck Index: 13,03702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a54fcee283659c6154da4eddffaef1718d046925&vid=8f1128967a01a32802f875d8c8d77de19679a735&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR