4-Aminobenzamidine dihydrochloride ≥98%

Synonyms: 4-Aminophenylamidine dihydrochloride, 4-Aminobenzene-1-carboximidamide dihydrochloride

857661-1G 857661-10G 857661-50G
220098-351EA 64.08 USD
220098-351 220098-350 220098-352
4-Aminobenzamidine dihydrochloride ≥98%
4-Aminobenzamidine dihydrochloride
Formula: C₇H₁₁Cl₂N₃
MW: 208.09 g/mol
Melting Pt: 290…291 °C
MDL Number: MFCD00013001
CAS Number: 2498-50-2
EINECS: 219-692-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR