Basic fuchsin, certified by the Biological Stain Commission

Danger

Synonyms: CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline

857343-25G 857343-100G
220098-340EA 190.4 USD
220098-340 220098-339
Basic fuchsin, certified by the Biological Stain Commission
Basic fuchsin
Formula: C₂₀H₂₀ClN₃
MW: 337.85 g/mol
Melting Pt: 235 °C
Density: 1.22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012569
CAS Number: 632-99-5
EINECS: 211-189-6
Merck Index: 12,05686
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ecca7a0bd05ed28f29bfcca62c8b34fddba95993&vid=a30faa9041a621f80fc892aa6aab88ac302082e9&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR