Stearoyl chloride, Technical Grade

85730-100ML 85730-500ML
220098-329EA 451.23 USD
220098-329 220098-330
Stearoyl chloride, Technical Grade
Stearoyl chloride
Formula: C₁₈H₃₅ClO
Boiling Pt: 348 °C (1013 hPa)
Melting Pt: 164…167 °C
Density: 0.9007 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000744
CAS Number: 112-76-5
EINECS: 204-004-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR