D-(-)-Leucine ≥99%

Synonyms: H-D-Leu-OH, (R)-(-)-Leucine

855448-2.5G 855448-25G 855448-10G
220097-012EA 70 USD
220097-012 220097-013 220097-011
D-(-)-Leucine ≥99%
D-(-)-Leucine
Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Boiling Pt: 226 °C (1013 hPa)
MDL Number: MFCD00063088
CAS Number: 328-38-1
EINECS: 206-327-7

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR