5-Bromouracil

Synonyms: 5-Bromopyrimidine-2,4(1H,3H)-dione, Bromouracil

852473-25G 852473-5G
220097-995EA 88.09 USD
220097-995 220097-996
5-Bromouracil
5-Bromouracil
Formula: C₄H₃BrN₂O₂
MW: 190.98 g/mol
Boiling Pt: 384 °C (1013 hPa)
Melting Pt: 165 °C
MDL Number: MFCD00006017
CAS Number: 51-20-7
EINECS: 200-084-0
Merck Index: 13,01425

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR