Silver diethyldithiocarbamate ≥99.0% ACS

Warning

Synonyms: N,N-Diethyldithiocarbamic acid silver salt, Diethyldithiocarbamic acid silver salt, DETC, (Diethyldithiocarbamato-S,S')silver

85180-10G 85180-50G
220097-547EA 109.23 USD
220097-547 220097-548
Silver diethyldithiocarbamate ≥99.0% ACS
Silver diethyldithiocarbamate
Formula: (C₂H₅)₂NCSSAg
MW: 256.14 g/mol
Melting Pt: 176…178.5 °C
MDL Number: MFCD00004929
CAS Number: 1470-61-7
EINECS: 216-003-7
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 99.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR