L(-)-Xylose mixture of anomers

Synonyms: (2S,3R,4S)-2,3,4,5-Tetrahydroxypentanal, L-(-)-Xylose

851590-25G 851590-5G
220097-541EA 58.58 USD
220097-541 220097-542
L(-)-Xylose mixture of anomers
L(-)-Xylose
Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00151096
CAS Number: 609-06-3
EINECS: 210-174-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR