2-Aminoisobutyric acid ≥98%

Synonyms: α-Aminoisobutyric acid, 2-Amino-2-methylpropionic acid, 2-Metilalanina, H-Aib-OH, 2-Methylalanine, 2-Amino-2-methylpropanoic acid

850993-25G 850993-100G
220097-973EA 64.51 USD
220097-973 220097-972
2-Aminoisobutyric acid ≥98%
2-Aminoisobutyric acid
Formula: C₄H₉NO₂
MW: 103.12 g/mol
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: 335 °C
Density: 1.278 g/cm³ (20 °C)
MDL Number: MFCD00008049
CAS Number: 62-57-7
EINECS: 200-544-0
Merck Index: 13,00444

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR