(3,4-Dihydroxyphenyl)acetic acid ≥98%

Warning

Synonyms: DOPAC, 3,4-Dihydroxybenzeneacetic acid, 3,4-Dihydroxyphenylacetic acid

850217-5G 850217-1G
220097-954EA 219.69 USD
220097-954 220097-953
(3,4-Dihydroxyphenyl)acetic acid ≥98%
(3,4-Dihydroxyphenyl)acetic acid
Formula: (C₆H₃(OH)₂)CH₂COOH
MW: 168.15 g/mol
Boiling Pt: 337 °C (1013 hPa)
Melting Pt: 131 - 132 °C
MDL Number: MFCD00004338
CAS Number: 102-32-9
EINECS: 203-024-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR