Dibutyl sebacate ≥97.0%

Warning

Synonyms: Sebacic acid dibutyl ester, 1,10-Decanedioic acid dibutyl ester, Dibutyl decanedioate

84840-1L 84840-250ML
220097-918EA 118.46 USD
220097-918 220097-919
Dibutyl sebacate ≥97.0%
Dibutyl sebacate
Formula: C₁₈H₃₄O₄
MW: 314.47 g/mol
Boiling Pt: 180 °C (1013 hPa)
Melting Pt: –11 °C
Density: 0.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00027218
CAS Number: 109-43-3
EINECS: 203-672-5

Order Now

Specification Test Results

Assay 97.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR