Sebacic acid ≥95.0% (by GC)

Synonyms: Decanedioic acid, 1,8-Octanedicarboxylic acid, 1,10-Decanedioic acid

84809-1KG 84809-100G
220097-911EA 103.38 USD
220097-911 220097-910
Sebacic acid ≥95.0% (by GC)
Sebacic acid
Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
Boiling Pt: 374 °C (1013 hPa)
Melting Pt: 127 - 131 °C
Density: 1.21 g/cm³ (20 °C)
Flash Pt: 221 °C
Storage Temperature: Ambient
MDL Number: MFCD00004440
CAS Number: 111-20-6
EINECS: 203-845-5
REACH: 01-2119519212-52
Merck Index: 12,08558

Order Now

Specification Test Results

Assay 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR