Ruthenium(III) chloride hydrate 38.0-42.0% (Ru basis)

Danger

Synonyms: Ruthenium(III) chloride xhydrate, Ruthenium trichloride hydrate

84050-1G-F 84050-5G-F
220097-822EA 48.46 USD
220097-822 220097-823
Ruthenium(III) chloride hydrate 38.0-42.0% (Ru basis)
Ruthenium(III) chloride hydrate
Formula: Cl₃Ru·xH₂O
MW: 207.43 g/mol (anhy)
Melting Pt: > 300 °C
Density: 3.11 g/cm³ (20 °C)
MDL Number: MFCD00149844
CAS Number: 14898-67-0
EINECS: 233-167-5
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay 38.0-42.0%
Ru basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR