Potassium methanesulfonate

83000-5G-F 83000-25G-F
220097-516EA 67.09 USD
220097-516 220097-515
Potassium methanesulfonate
Potassium methanesulphonate
Formula: CH₃KO₃S
MDL Number: MFCD00070544
CAS Number: 2386-56-3
EINECS: 219-202-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR