Polyethylene glycol 20,000

Synonyms: PEG 20000, Ethylene glycol polymer 20000, PEG 20,000

81300-1KG 81300-5KG
220097-077EA 130.77 USD
220097-077 220097-078
Polyethylene glycol 20,000
Polyethylene glycol 20,000
Formula: H(OCH₂CH₂)nOH
MW: 20000 g/mol
Melting Pt: 58 - 63 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR