Hexachloroplatinic(IV) acid hydrate

Danger

Synonyms: Hydrogen hexachloroplatinate (IV) hydrate, Hydrogen hexachloroplatinate hydrate, Chloroplatinic acid hydrate, Dihydrogen hexachloroplatinate (IV) hydrate, Hydrogen hexachloroplatinate(IV) hydrate

81080-1G 81080-5G
220096-952EA 86.54 USD
220096-952 220096-953
Hexachloroplatinic(IV) acid hydrate
Hexachloroplatinic(IV) acid hydrate
Formula: H₂Cl₆Pt·H₂O
MW: 427.83 g/mol
Melting Pt: 60 °C
MDL Number: MFCD00149909
CAS Number: 26023-84-7
EINECS: 241-010-7
UN: 2507
ADR: 8,III

Order Now

Specification Test Results

Assay ~38% Pt basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR