(R)-(-)-1-Phenyl-2-propanol

78926-1ML 78926-5ML
220096-641EA 1255.38 USD
220096-641 220096-160
(R)-(-)-1-Phenyl-2-propanol
(R)-(-)-1-Phenyl-2-propanol
Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 114 °C (25 mmHg)
Density: 0.993 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 1572-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR