m-Phenylenediamine dihydrochloride

Danger

Synonyms: Benzene-1,3-diamine dihydrochloride, 1,3-Diaminobenzene dihydrochloride, 1,3-Phenylenediamine dihydrochloride, 1,3-Phenylenediammonium dichloride

78450-100G 78450-500G
220096-571EA 264.92 USD
220096-571 220096-572
m-Phenylenediamine dihydrochloride
m-Phenylenediamine dihydrochloride
Formula: C₆H₁₀Cl₂N₂
MW: 181.06 g/mol
Melting Pt: 277 - 278 °C
Storage Temperature: Ambient
MDL Number: MFCD00012975
CAS Number: 541-69-5
EINECS: 208-790-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR