1-Butyl-3-methylimidazoliumbis(trifluoromethylsulfonyl)imide

Synonyms: 1-Butyl-3-methylimidazolium Bis(trifluoromethanesulphonyl)imide, 1-Butyl-3-methyl-1H-imidazol-3-ium bis((trifluoromethyl)sulphonyl)amide

77896-5G-F 77896-1G-F
220096-485EA 176 USD
220096-485 220096-484
1-Butyl-3-methylimidazoliumbis(trifluoromethylsulfonyl)imide
1-Butyl-3-methylimidazoliumbis(trifluoromethylsulphonyl)imide
Formula: C₁₀H₁₅F₆N₃O₄S₂
MW: 419.37 g/mol
MDL Number: MFCD05664714
CAS Number: 174899-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR