1,10-Phenanthroline hydrochloride monohydrate

Danger

Synonyms: 1,10-Phenanthroline Chloride Monohydrate, 1,10-Phenanthrolinium chloride monohydrate, 1,10-Phenanthroline monohydrochloride monohydrate

77510-5G 77510-25G
220096-421EA 68.71 USD
220096-421 220096-420
1,10-Phenanthroline hydrochloride monohydrate
1,10-Phenanthroline hydrochloride monohydrate
Formula: C₁₂H₉ClN₂·1H₂O
MW: 234.68 g/mol
Melting Pt: 224 - 226 °C
Density: 1.385 g/cm³ (-100.15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150061
CAS Number: 18851-33-7
EINECS: 223-325-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99% (AT)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR