Potassium heptadecafluorooctane-1-sulfonate ≥98.0% (by titrimetric analysis)

Synonyms: Potassium perfluorooctanesulphonate, Heptadecafluoro-1-octanesulphonic acid potassium salt, Potassium perfluorooctane-1-sulphonate, Perfluoro-1-octanesulphonic acid potassium salt, Perfluorooctanesulphonic acid, potassium salt, Heptadecafluorooctanesulphonic acid potassium salt

77282-10G 77282-50G
220096-358EA 128.08 USD
220096-358 220096-359
Potassium heptadecafluorooctane-1-sulfonate ≥98.0% (by titrimetric analysis)
Potassium heptadecafluorooctane-1-sulphonate
Formula: C₈F₁₇KO₃S
MW: 538.22 g/mol
MDL Number: MFCD00066407
CAS Number: 2795-39-3
EINECS: 220-527-1

Order Now

Specification Test Results

Assay 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR