1-Pentanesulfonic acid sodium salt monohydrate ≥98.0% (by titrimetric analysis)

Synonyms: Sodium 1-pentanesulphonate monohydrate

76955-50G 76955-10G
220096-110EA 402.77 USD
220096-110 220096-109
1-Pentanesulfonic acid sodium salt monohydrate ≥98.0% (by titrimetric analysis)
1-Pentanesulphonic acid sodium salt monohydrate
Formula: C₅H₁₁NaO₃S·1H₂O
MW: 192.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007541
CAS Number: 207605-40-1
EINECS: 245-208-4

Order Now

Specification Test Results

Assay 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR