1-(4-Pyridyl)piperazine

Synonyms: 4-(1-Piperazinyl)pyridine, 1-(Pyridin-4-yl)piperazine

76763-5G-F 76763-1G-F
220096-087EA 183.08 USD
220096-087 220096-086
1-(4-Pyridyl)piperazine
1-(4-Pyridyl)piperazine
Formula: C₉H₁₃N₃
MW: 163.22 g/mol
Boiling Pt: 195 - 200 °C (0.3 -rr)
Melting Pt: 140 °C
MDL Number: MFCD00040745
CAS Number: 1008-91-9
EINECS: 213-764-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR