1-Butyl-3-methylimidazoliumtrifluoromethanesulfonate

Synonyms: 1-Butyl-3-methylimidazolium Trifluoromethanesulphonate

76420-25G-F 76420-5G-F
220095-581EA 100 USD
220095-581 220095-582
1-Butyl-3-methylimidazoliumtrifluoromethanesulfonate
1-Butyl-3-methylimidazoliumtrifluoromethanesulphonate
Formula: C₉H₁₅F₃N₂O₃S
MW: 288.29 g/mol
MDL Number: MFCD03427620
CAS Number: 174899-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR