Palladium(II) nitrate dihydrate

Danger

76070-5G 76070-1G
220095-947EA 576.92 USD
220095-947 220095-946
Palladium(II) nitrate dihydrate
Palladium(II) nitrate dihydrate
Formula: N₂O₆Pd·2H₂O
CAS Number: 32916-07-7
EINECS: 233-265-8
UN: 1477
ADR: 5.1,II

Order Now

Specification Test Results

Assay ~40% Pd basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR